Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Salicylate, Powder, USP, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18604103 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
75 g
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-604-103

Supplier: Spectrum Chemical Manufacturing Corporation MA128125GM

Only null left
Add to Cart
Add to Cart

MA128, 18917-95-8, C14H10MgO6·4H2O

Spectrum™ Chemical Magnesium Salicylate, Powder, USP is a non-steroidal anti-inflammatory drug (NSAID) in a group of drugs called salicylates. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 18917-95-8
Molecular Formula C14H18MgO10
Molecular Weight (g/mol) 370.59
MDL Number MFCD01766176,MFCD00045815
InChI Key NBQBEWAYWAMLJJ-UHFFFAOYSA-L
IUPAC Name magnesium(2+) bis(2-hydroxybenzoate) tetrahydrate
SMILES O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 103%
Grade USP
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.