Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium silicate, activated, for chromatography, 60-100 mesh
Click to view available options
Quantity:
250 g
1000 g
Description
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Solubility
Insoluble in water.
Notes
Incompatible with strong acids and hydrogen fluoride.
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Solubility
Insoluble in water.
Notes
Incompatible with strong acids and hydrogen fluoride.

Specifications
Specifications
Quantity | 1000 g |
Linear Formula | Mg2SiO4 |
Solubility Information | Insoluble in water. |
Formula Weight | 140.71 |
Chemical Name or Material | Magnesium silicate, activated, for chromatography |
Safety and Handling
EINECSNumber | 215-681-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction