Learn More
Magnesium silicate, activated, for chromatography, 60-100 mesh
Description
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Solubility
Insoluble in water.
Notes
Incompatible with strong acids and hydrogen fluoride.
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | Mg2SiO4 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 140.71 |
| Chemical Name or Material | Magnesium silicate, activated, for chromatography |
Safety and Handling
| EINECSNumber | 215-681-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.