Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium silicate, activated, for chromatography, 60-100 mesh

Catalog No. p-7026481
Click to view available options
Quantity:
250 g
1000 g

CAS: 1343-88-0 | MgO3Si | 100.39 g/mol

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids and hydrogen fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Synonym Activated magnesium silicate

Specifications

Quantity 250 g
Linear Formula Mg2SiO4
Solubility Information Insoluble in water.
Formula Weight 140.71
Chemical Name or Material Magnesium silicate, activated, for chromatography
EINECSNumber 215-681-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.