Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate, 99%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
7487-88-9
MgO4S
120.37
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]
7487-88-9
MgO4S
120.37
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula MgSO4
Merck Index 15,5755
Solubility Information Solubility in water: 26g/100mL (0°C).
Formula Weight 120.37
Percent Purity 99%
Grade Analysis
Chloride 50ppm max
Packaging Plastic bottle
Solubility Solubility in water: 26g/100mL (0°C).
Melting Point 1124.0°C
Quantity 250g
Chemical Name or Material Magnesium sulfate
Show More Show Less

EINECSNumber : 231-298-2