Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate, 99%, extra pure, dried, contains 3 to 4 moles of water, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic Bottle
Plastic drum
22189-08-8
MgO4S·xH2O
120.36
MFCD00149787
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]
22189-08-8
MgO4S·xH2O
120.36
MFCD00149787
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate,magnesium sulphate,magnesium sulfate anhydrous,sulfuric acid magnesium salt 1:1,bitter salt,sal angalis,magnesium sulfate 1:1,mgso4,sal de sedlitz,tomix ot
24083
CHEBI:32599
magnesium;sulfate
[O-]S(=O)(=O)[O-].[Mg+2]
Adsorbent
Acidity Alkalinity Passes Test
Assay Percent Range 98.5% min. (On dried substance) (Complexometry)
Linear Formula MgSO4·xH2O
Arsenic (As) 2ppm max.
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
Formula Weight 120.36
Physical Form Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Chloride 400ppm max
Chlorine (Cl) 400ppm max.
Density 2.6600g/mL
Heavy Metals (as Pb) 20ppm max.
Iron (Fe) 10ppm max.
Loss on Drying 26% to 32% (400 to 500 °C)
Packaging Plastic Bottle
Solubility Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
Specific Gravity 2.66
Color White
Melting Point 1124.0°C
Quantity 1kg
Chemical Name or Material Magnesium sulfate, Dried, contains 3 to 4 Moles of Water
Show More Show Less