Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate, anhydrous, 99.5% min

Catalog No. aa3333736
Click to view available options
Quantity:
500 g
2 kg
10 kg

CAS: 7487-88-9 | MgO4S | 120.36 g/mol

Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124°C (decomposition)
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5% min
Solubility Information Soluble in water. Slightly soluble in alcohol,glycerol. Insoluble in acetone.
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.37
Sensitivity Hygroscopic
Density 2.66 g/mL
Chemical Name or Material Magnesium sulfate, anhydrous, anhydrous
EINECSNumber 231-298-2
RTECSNumber OM4500000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.