Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Sulfate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
125 g
500 g
2.5 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Magnesium Sulfate, Anhydrous, USP is the raw material for a common magnesium-based, pharmaceutical preparation called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it′s anhydrous form, magnesium sulfate is used as a drying agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..
Specifications
Specifications
pH | 5.0-9.2 |
CAS Max % | 100% |
Quantity | 50 kg |
Percent Purity | 99 to 100.5% |
Grade | USP |
Loss on Drying | 2% |
Packaging | Fiber Drum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction