Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Sulfate Heptahydrate, 27% w/v Volumetric Solution, Reagents
Click to view available options
:
Quantity: 1 L
Specifications
Specifications
Chemical Name or Material | Magnesium Sulfate Heptahydrate, 27% w/v Volumetric Solution |
Boiling Point | 101°C |
CAS | 7732-18-5, 10034-99-8 |
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Percent Purity | 27% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.1083 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction