Learn More
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%
Description
Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Crystals |
Quantity | 2 kg |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Soluble in in water and acid. Slightly soluble in alcohol and glycerol. |
Formula Weight | 246.48 (120.37 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Appearance | White crystals |
Density | 1.68 |
Show More |
Safety and Handling
EINECSNumber | 231-298-2 |
RTECSNumber | OM4508000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.