Learn More
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%
Description
Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystals |
| Quantity | 500 g |
| Assay Percent Range | 98.0-102.0% |
| Solubility Information | Soluble in in water and acid. Slightly soluble in alcohol and glycerol. |
| Formula Weight | 246.48 (120.37 Anhydrous) |
| Grade | ACS |
| Odor | Odorless |
| Appearance | White crystals |
| Density | 1.68 g/cm3 at 20°C |
| Show More |
Safety and Handling
| EINECSNumber | 231-298-2 |
| RTECSNumber | OM4508000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.