Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Sulfate Heptahydrate (Crystalline/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Specifications
Specifications
Melting Point | 770°C |
Color | White |
pH | 5 to 8.2 |
Physical Form | Solid |
Quantity | 50 kg |
Assay Percent Range | 98 to 102 % |
Formula Weight | 246.47 |
Percent Purity | 98 to 102% |
Grade | Certified ACS |
Identification | Pass Test |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction