Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Sulfate Heptahydrate, Purified (Epsom Salts), Reagents
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Specifications
Specifications
| Chemical Name or Material | Magnesium Sulfate Heptahydrate, Purified (Epsom Salts) |
| Boiling Point | 200°C |
| Color | Colorless/White |
| CAS | 10034-99-8 |
| Physical Form | Solid |
| Quantity | 12 kg |
| Molecular Formula | MgSO4, 7H2O |
| Molecular Weight (g/mol) | 246.4746 |
| Grade | Pure |
| Packaging | Poly Pail |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction