Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate hydrate, Puratronic™, 99.997% (metals basis)

Catalog No. p-7048532
Click to view available options
Quantity:
10 g
50 g

CAS: 22189-08-8 | MgO4S | 120.36 g/mol

Magnesium sulfate is used as an anticonvulsant, a cathartic, and an electrolyte replenisher in the treatment of pre-eclampsia and eclampsia. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 10 g
Merck Index 14,5691
Solubility Information Soluble in water (360mg/ml at 20°C),ether (11.6mg/ml at 18°C),alcohol,and glycerin. Insoluble in acetone.
Formula Weight 120.37 (Anhydrous)
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 1.68 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium sulfate hydrate
Show More Show Less
EINECSNumber 231-298-2
RTECSNumber OM4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.