Alfa Aesar™ Magnesium sulfate hydrate, Puratronic™, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 1080109

Quantity 10g

 View more versions of this product

Catalog No. AA1080109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22189-08-8
    Formula Weight 120.37 (Anhydrous)
    MDL Number MFCD00149787
    Merck Index 14,5691
    Chemical Name or Material Magnesium sulfate hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 120.37 (Anhydrous)
    TSCA TSCA
    Physical Form Crystalline Aggregates
    Merck Index 14,5691
    Color White
    Quantity 10g
    Density 1.68g/mL
    Odor Odorless
    MDL Number MFCD00149787
    CAS 22189-08-8
    RTECS Number OM4500000
    Chemical Name or Material Magnesium sulfate hydrate
    Assay 99.997%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit