Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Sulfate Solution, 250 g/L in 2.0 N Sulfuric Acid, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Chemical Name or Material | Magnesium Sulfate Solution |
| Color | Colorless |
| CAS | 7487-88-9, 10034-99-8, 7664-93-9 |
| CAS Min % | 73.0, 18.14, 7.34 |
| CAS Max % | 75.0, 18.88, 7.64 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | 2.0 N Sulfuric Acid |
| pH | <1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction