Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium tert-butoxide, 93+%
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White to Gray |
Assay Percent Range | 93+% |
Packaging | Plastic bottle |
Quantity | 250 g |
Solubility Information | Solubility in water: reacts violently |
Formula Weight | 170.53 |
Percent Purity | 93+% |
Physical Form | Powder |
Chemical Name or Material | Magnesium tert-butoxide |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if inhaled. Reacts violently with water. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present |
EINECSNumber | 250-931-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction