Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium tetraborate, 99%

Catalog No. AA4389214
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA4389214

Supplier: Thermo Scientific Chemicals 04389214

Only null left
Add to Cart

CAS: 12007-62-4 | B2Mg3O6 | 190.529 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-62-4
Molecular Formula B2Mg3O6
Molecular Weight (g/mol) 190.529
MDL Number MFCD02688001
InChI Key NFMWFGXCDDYTEG-UHFFFAOYSA-N
Synonym acmc-1bvay, boric acid h3bo3 , magnesium salt 2:3, ksc171o6l, trimagnesium 2+ diborate, parent, trimagnesium 2+ ion diborate
PubChem CID 21981398
IUPAC Name trimagnesium;diborate
SMILES B([O-])([O-])[O-].B([O-])([O-])[O-].[Mg+2].[Mg+2].[Mg+2]

Specifications

Physical Form Powder
Quantity 25 g
Formula Weight 179.54
Percent Purity 99%
Odor Odorless
Chemical Name or Material Magnesium tetraborate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.