Alfa Aesar™ Magnesium thiosulfate hexahydrate, 99%

Manufacturer: Alfa Aesar™ 4611436

Quantity 500g

 View more versions of this product

Catalog No. AA4611436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13446-30-5
    Molecular Formula MgO3S2·6H2O
    Linear Formula MgS2O3·6H2O
    Formula Weight 244.53 (136.44 Anhydrous)
    MDL Number MFCD00169871
    Merck Index 14,5694
    Synonym Magnesium hyposulfite
    Chemical Name or Material Magnesium thiosulfate hexahydrate

    Specifications

    Formula Weight 244.53 (136.44 Anhydrous)
    Melting Point 170°C (in -3H2O)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula MgO3S2·6H2O
    Linear Formula MgS2O3·6H2O
    Synonym Magnesium hyposulfite
    TSCA TSCA
    Physical Form Crystalline
    CAS 13446-30-5
    Merck Index 14,5694
    Color White
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00169871
    Solubility Information Soluble in water; Insoluble in alcohol
    Health Hazard 1 Warning
    Quantity 500g
    Density 1.818g/mL
    RTECS Number OM4610000
    Chemical Name or Material Magnesium thiosulfate hexahydrate
    Assay 99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit