Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Magnesium thiosulfate hexahydrate, 99%

Manufacturer:  Alfa Aesar™ 4611422

Quantity 100g

 View more versions of this product

Catalog No. AA4611422


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13446-30-5
MgS2O3·6H2O
MFCD00169871
Magnesium hyposulfite
MgO3S2·6H2O
244.53 (136.44 Anhydrous)
14,5694
Magnesium thiosulfate hexahydrate

Specifications

13446-30-5
MgO3S2·6H2O
244.53 (136.44 Anhydrous)
14,5694
Soluble in water; Insoluble in alcohol
White
100g
99%
Crystalline
MgS2O3·6H2O
MFCD00169871
Magnesium hyposulfite
1.818g/mL
170°C (in -3H2O)
Magnesium thiosulfate hexahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OM4610000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit