Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Magnetic Drape Holders
SCI_ED

For use with magnetic drapes for boards equipped with Magnetic Base System.

$313.00

Specifications

Type Magnetic Drape Holder
For Use With (Equipment) Boards equipped with Magnetic Base System
View More Specs

Products 2
Catalog Number Mfr. No. Size Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Diameter (Metric) Price Quantity & Availability  
14-370-228
SCI_ED
View Documents
AIMS™
GDHL
Large 2.5 cm
Each for $313.00
Only null left
Add to Cart
 
14-370-229
SCI_ED
View Documents
AIMS™
GDHS
Small 1.6 cm
Each for $313.00
Only null left
Add to Cart
 
Description

Description

  • Available in small and large sizes
  • Set of 4
Specifications

Specifications

Magnetic Drape Holder
Boards equipped with Magnetic Base System
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.