Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Magnetic Screw Thread Caps
SDP

Catalog No. 06611976 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Color:
Blue (Septa)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
06-611-976 Blue (Septa)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-976 Supplier Restek™ Supplier No. 23091
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Magnetic caps and septa for SPME

  • 18 mm magnetic screw-thread headspace caps
  • Blue PTFE/silicone
  • 1.5 mm thick septa for SPME

Specifications

Product Type Threaded Closure
Description Screw-thread caps size 18-425
Material PTFE, Silicone
Color Blue (Septa)
Quantity 1000 Pk.
Thickness 1.5 mm (septa)
For Use With (Equipment) Autosampler
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.