Learn More
Eisco™ Magnetite Mineral Specimen
SCI_ED
Catalog No. S24698 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24698 12/Pk. 2 to 3 cm
S24697 1 Ea. 2 to 3 cm
S26613 10/Pk. 2 to 3 cm
S26673 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24698 Supplier Eisco™ Supplier No. ESNG0020PK12
Add to Cart
Add to Cart
Magnetite grains align themselves with the earth's magnetic field, preserving a record of the field's direction when forming igneous rocks.
  • Magnetite's attraction to magnets and distinctive black streak make it simple to identify
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Strongly magnetic (will be attracted to magnets, but cannot be used to attract other metallic objects)
Specifications
Product Type Mineral
For Use With (Application) Mineral identification exercises
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Magnetite