Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnolol

Catalog No. AC465510010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465510010 1 g
AC465512500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465510010 Supplier Thermo Scientific Chemicals Supplier No. 465510010
Add to Cart

CAS: 528-43-8 | C18H18O2 | 266.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-43-8
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
InChI Key VVOAZFWZEDHOOU-UHFFFAOYSA-N
IUPAC Name 5,5'-bis(prop-2-en-1-yl)-[1,1'-biphenyl]-2,2'-diol
SMILES OC1=CC=C(CC=C)C=C1C1=CC(CC=C)=CC=C1O

Specifications

Melting Point 101.0°C to 102.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in ethanol
Formula Weight 266.34
Chemical Name or Material Magnolol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye damage.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.