Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGOH Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297072JF549
Description
MAGOH Polyclonal antibody specifically detects MAGOH in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
MAGOH | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
MAGOHAMAGOH1, mago-nashi (Drosophila) homolog, proliferation-associated, mago-nashi homolog, proliferation-associated (Drosophila), protein mago nashi homolog | |
Rabbits immunized with purified, recombinant Human MAGOH (Accession#: NP_002361.1, Met1-Ile146) | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
4116 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction