Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGOH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118043 Supplier Invitrogen™ Supplier No. PA5118043
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGOH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOH
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene MAGOH
Gene Accession No. P61326
Gene Alias ch211-216k22.5; hypothetical protein LOC550395; mago homolog, exon junction complex core component; mago homolog, exon junction complex subunit; mago nashi; Magoh; MAGOH1; MAGOHA; Mago-m; mago-nashi homolog, proliferation-associated; mago-nashi homolog, proliferation-associated (Drosophila); mago-nashi-like proliferation-associated protein; Mos2; Protein mago nashi homolog; RP5-1024G6.4; zgc:112220
Gene Symbols MAGOH
Host Species Rabbit
Immunogen E. coli-derived Human MAGOH fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4116
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.