Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGOH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560208 Supplier Invitrogen™ Supplier No. PA560208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLRYANNSNY KNDVMIRKEA YVHKSVMEEL KRIIDDSEIT KEDDALWPPP DRVGRQELEI VIGDEHIS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGOH
Gene Accession No. P61326
Gene Alias ch211-216k22.5; hypothetical protein LOC550395; mago homolog, exon junction complex core component; mago homolog, exon junction complex subunit; mago nashi; Magoh; MAGOH1; MAGOHA; Mago-m; mago-nashi homolog, proliferation-associated; mago-nashi homolog, proliferation-associated (Drosophila); mago-nashi-like proliferation-associated protein; Mos2; Protein mago nashi homolog; RP5-1024G6.4; zgc:112220
Gene Symbols MAGOH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGOH. Recombinant protein control fragment (Product #RP-104351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.