Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGOH Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981750
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1750 Supplier Bioss Supplier No. BS11882R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.

Specifications

Antigen MAGOH
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAGOH
Gene Accession No. P61326
Gene Alias BOS_3517; ch211-216k22.5; hypothetical protein LOC550395; LOC100354873; mago homolog, exon junction complex core component; mago homolog, exon junction complex subunit; mago nashi; magoh; MAGOH1; MAGOHA; Mago-m; mago-nashi homolog; mago-nashi homolog, proliferation-associated; mago-nashi homolog, proliferation-associated (Drosophila); Mago-nashi-like proliferation-associated protein; Mos2; Protein mago nashi homolog; RP5-1024G6.4; zgc:112220
Gene Symbols MAGOH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAGOH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4116
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.