Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGOH Recombinant Rabbit Monoclonal Antibody (3J7K2)
GREENER_CHOICE

Catalog No. PIMA542825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542825 Supplier Invitrogen™ Supplier No. MA542825
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, A-549, Jurkat, Mouse spleen, Rat thymus, Rat lung, Rat spleen. Immunogen sequence: SEITKEDDAL WPPPDRVGRQ ELEIVIGDEH ISFTTSKIGS LIDVNQSKDP EGLRVFYYLV QDLKCLVFSL IGLHFKIKPI.

Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOH
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3J7K2
Concentration 2.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAGOH
Gene Accession No. P61326, P61327, Q27W02
Gene Alias ch211-216k22.5; hypothetical protein LOC550395; mago homolog, exon junction complex core component; mago homolog, exon junction complex subunit; mago nashi; Magoh; MAGOH1; MAGOHA; Mago-m; mago-nashi homolog, proliferation-associated; mago-nashi homolog, proliferation-associated (Drosophila); mago-nashi-like proliferation-associated protein; Mos2; Protein mago nashi homolog; RP5-1024G6.4; zgc:112220
Gene Symbols MAGOH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 67-146 of human MAGOH (P61326).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17149, 298385, 4116
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.