Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGOHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570585 Supplier Thermo Scientific Supplier No. PA570585
Add to Cart
Add to Cart

MAGOHB Polyclonal Antibody

MAGOHB Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOHB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAGOHB
Gene Accession No. Q96A72
Gene Alias 2010012C16Rik; LOC100063178; mago; mago homolog B, exon junction complex core component; mago homolog B, exon junction complex subunit; mago nashi protein; Magoh; MAGOH2; Magohb; Magoh-rs1; mago-nashi homolog 2; mago-nashi homolog B; mago-nashi homolog B (Drosophila); MGN2; protein mago nashi homolog; protein mago nashi homolog 1-related; protein mago nashi homolog 2; protein mago nashi homolog 2; protein mago nashi homolog
Gene Symbols MAGOHB
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MAGOHB (aa 65-114).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55110
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.