Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGOHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116666 Supplier Invitrogen™ Supplier No. PA5116666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Jurkat, PC-12, U-87MG Immunogen sequence: MAVASDFYLR YYVGHKGKFG HEFLEFEFRP DGKLRYANNS NYKNDVMIRK EAYVHKSVME ELKRIIDDSE ITKEDDALWP PPDRVGRQEL EIVIGDEHIS FTTSKIGSLI DVNQSKDPEG LRVFYYLVQD LKCLVFSLIG LHFKIKPI.

MAGOHB is a protein coding gene involved in mRNA splicing and in the nonsense-mediated decay (NMD) pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOHB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAGOHB
Gene Accession No. Q96A72
Gene Alias 2010012C16Rik; LOC100063178; mago; mago homolog B, exon junction complex core component; mago homolog B, exon junction complex subunit; mago nashi protein; Magoh; MAGOH2; Magohb; Magoh-rs1; mago-nashi homolog 2; mago-nashi homolog B; mago-nashi homolog B (Drosophila); MGN2; protein mago nashi homolog; protein mago nashi homolog 1-related; protein mago nashi homolog 2; protein mago nashi homolog 2; protein mago nashi homolog
Gene Symbols MAGOHB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human MAGOHB (NP_0605181).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55110, 690303
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.