Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGP-2/MFAP5 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300035MFV610
Description
MAGP-2/MFAP5 Polyclonal antibody specifically detects MAGP-2/MFAP5 in Human samples. It is validated for ELISASpecifications
MAGP-2/MFAP5 | |
Polyclonal | |
ELISA | |
MAGP-2, MAGP2MFAP-5, Microfibril-associated glycoprotein 2, microfibrillar associated protein 5, microfibrillar-associated protein 5, MP25microfibril-associated glycoprotein-2 | |
Produced in rabbits immunized with purified, recombinant Human MAGP-2/MFAP5 (Uniprot#: Q13361; Met1-Leu173) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
8076 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction