Learn More
Cytiva MagRack 6
Catalog No. 45002170 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
45-002-170 1 Ea.
1 options
Catalog No. 45-002-170 Supplier Cytiva Supplier No. 28948964
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Use for small-scale sample preparation using magnetic beads. Cytiva Life Sciences™ MagRack 6 enables handling of up to six samples in 1.5 ml microcentrifuge tubes.
When tubes are placed in the rack, the magnetic beads are attracted to the magnet within a few seconds. Tubes can be locked in position and the rack inverted, which allows easy removal of waste solutions between steps in the protocol, while the magnetic beads remain in the tube.
The rack consists of anodized aluminum housing with a detachable plastic magnetic bar containing six neodymium magnets.
  • Rapid and simple processing for Mag Sepharose™ purification and enrichment applications
  • No need for centrifugation
  • Vary the amount of beads to meet specific requirements
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Application) Protein Sample Preparation
Product Type MagRack 6