Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544929 Supplier Invitrogen™ Supplier No. PA544929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ARWRFWCVSV TMVVALLIVC DVPSASAQRK KEMVLSEKVS QLMEWTNKRP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 91%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

MAGT1 encodes a magnesium cation transporter protein that localizes to the cell membrane. This protein also associates with N-oligosaccharyl transferase and therefore may have a role in N-glycosylation. Mutations in this gene cause mental retardation X-linked type 95 (MRX95). This gene may have multiple in-frame translation initiation sites, one of which would encode a shorter protein with an N-terminus containing a signal peptide at amino acids 1-29.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Magt1
Gene Accession No. Q9H0U3
Gene Alias 2410001C15Rik; 2610529C04Rik; 2810482I07Rik; bA217H1.1; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit MAGT1; Iag2; IAP; Implantation-associated protein; magnesium transporter 1; magnesium transporter protein 1; MAGT1; MRX95; Oligosaccharyl transferase subunit MAGT1; oligosaccharyltransferase 3 homolog B; OST3B; PRO0756; PSEC0084; UNQ628/PRO1244; XMEN
Gene Symbols Magt1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RP11-217H1.1 (aa 3-52).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84061
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.