Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAK Antibody (OTI5A7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272580AF405
Description
MAK Monoclonal specifically detects MAK in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAK | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAK | |
Recombinant protein fragment corresponding to amino acids 291-623 of human MAK (NP_005897) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5A7 | |
Western Blot, Immunohistochemistry | |
dJ417M14.2, EC 2.7.11, EC 2.7.11.22, male germ cell-associated kinaseserine/threonine protein kinase MAK, serine/threonine-protein kinase MAK | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
4117.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction