Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAK Antibody (OTI5A7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272580
Description
MAK Monoclonal specifically detects MAK in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAK | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
dJ417M14.2, EC 2.7.11, EC 2.7.11.22, male germ cell-associated kinaseserine/threonine protein kinase MAK, serine/threonine-protein kinase MAK | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
4117 | |
Human, Rat | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI5A7 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MAK | |
Human recombinant protein fragment corresponding to amino acids 291-623 of human MAK(NP_005897) produced in E.coli. | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction