Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAK Monoclonal Antibody (OTI1G2)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527054

Catalog No. PIMA527054


Only null left
Add to Cart

Description

Description

MAK Monoclonal Antibody for Western Blot, IHC (P)

The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P20794
MAK
Human recombinant protein fragment corresponding to amino acids 291-623 of MAK produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1G2
Unconjugated
MAK
A930010O05Rik; dJ417M14.2; MAK; male germ cell associated kinase; male germ cell-associated kinase; male germ cell-associated kinase retinal-enriched isoform; protein kinase RCK; Rck; RP62; serine/threonine protein kinase MAK; serine/threonine-protein kinase MAK; testicular secretory protein Li 28; Unknown (protein for MGC:134523)
Mouse
Affinity chromatography
RUO
4117
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.