Learn More
MAK Mouse anti-Human, Clone: OTI4H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808784

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized.Specifications
MAK | |
Monoclonal | |
Unconjugated | |
MAK | |
RP62 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4117 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P20794 | |
MAK | |
Human recombit protein fragment corresponding to amino acids 291-623 of human MAK produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.