Learn More
Invitrogen™ MAK Polyclonal Antibody

Description
MAK Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | MAK |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | MAK |
| Gene Accession No. | P20794, Q04859 |
| Gene Alias | A930010O05Rik; dJ417M14.2; MAK; male germ cell associated kinase; male germ cell-associated kinase; male germ cell-associated kinase retinal-enriched isoform; protein kinase RCK; Rck; RP62; serine/threonine protein kinase MAK; serine/threonine-protein kinase MAK; testicular secretory protein Li 28; Unknown (protein for MGC:134523) |
| Gene Symbols | MAK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.