Learn More
Invitrogen™ MAK10 Polyclonal Antibody

Description
A suggested positive control is rat heart tissue lysate. PA5-20783 can be used with blocking peptide PEP-0897.
Specifications
Specifications
| Antigen | MAK10 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NAA35 |
| Gene Accession No. | Q5VZE5, Q6DKG0, Q6PHQ8 |
| Gene Alias | A330021G12Rik; A330027C19Rik; AI158944; bA379P1.1; C030004C14Rik; corneal wound healing related protein; corneal wound healing-related protein; corneal wound-healing-related protein; EGAP; Emb8; Embryonic growth-associated protein; embryonic growth-associated protein homolog; Mak10; MAK10 homolog, amino-acid N-acetyltransferase subunit; MAK10P; N(alpha)-acetyltransferase 35, NatC auxiliary subunit; NAA35; Nalpha acetyltransferase 35; N-alpha-acetyltransferase 35, NatC auxiliary subunit; protein MAK10 homolog; RAT52; RP11-379P1.1; T4a |
| Gene Symbols | NAA35 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.