Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAK16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560704
Description
Immunogen sequence: QSDDVIWDTL GNKQFCSFKI RTKTQSFCRN EYSLTGLCNR SSCPLANSQY ATIKEEKGQC YLYMKV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
MAK16 gene ontology annotations related to this gene include RNA binding.
Specifications
MAK16 | |
Polyclonal | |
Unconjugated | |
Mak16 | |
2600016B03Rik; AI314911; MAK16; MAK16 homolog; MAK16 homolog (S. cerevisiae); MAK16L; NNP78; Protein MAK16 homolog; Protein RBM13; Rbm13; RGD1311297; RNA binding motif protein 13; RNA binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84549 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXY0 | |
Mak16 | |
Recombinant protein corresponding to Human MAK16. Recombinant protein control fragment (Product #RP-93955). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction