Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAK16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562217
Description
Immunogen sequence: FIRHKCKQRF TKITQYLIRI RKLTLKRQRK LVPLSKKVER REKRREEKAL IAAQLDNAIE KELLERLKQD TYGDIYNFPI HAFDKALEQQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
MAK16 gene ontology annotations related to this gene include RNA binding.
Specifications
MAK16 | |
Polyclonal | |
Unconjugated | |
Mak16 | |
2600016B03Rik; AI314911; MAK16; MAK16 homolog; MAK16 homolog (S. cerevisiae); MAK16L; NNP78; Protein MAK16 homolog; Protein RBM13; Rbm13; RGD1311297; RNA binding motif protein 13; RNA binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84549 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXY0 | |
Mak16 | |
Recombinant protein corresponding to Human MAK16. Recombinant protein control fragment (Product #RP-93956). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction