Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Makerspace 3-5 Classroom Refill
SCI_ED

Catalog No. S02756
Click to view available options
Class Size:
1 Class

Catalog No. S02756

Supplier: Hand2Mind 92929

Only null left
Add to Cart
Add to Cart

Foster creativity and innovation with engineering concepts using our new Makerspace Solutions. They're an affordable, fully loaded, makerspace solution for classrooms, STEM labs, and libraries.

  • Foster creativity and innovation with engineering concepts using our new Makerspace Solutions.
  • They're an affordable, fully loaded, makerspace solution for classrooms, STEM labs, and libraries.

Specifications

Grade Level Elementary
Grade 3 to 5
Class Size 1 Class
Product Type Makerspace Classroom Refill
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.