Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Malachite Green

Catalog No. AC611255000
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 569-64-2 | C23H25ClN2 | 364.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-64-2
Molecular Formula C23H25ClN2
Molecular Weight (g/mol) 364.92
MDL Number MFCD00011751
InChI Key FDZZZRQASAIRJF-UHFFFAOYSA-M
PubChem CID 11294
ChEBI CHEBI:72449
IUPAC Name bis[4-(dimethylamino)phenyl](phenyl)methylium chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Beilstein 13, 744
Merck Index 15, 5763
Identification Pass Test
Formula Weight 364.92
Quantity 500 g
Chemical Name or Material Malachite Green

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.