Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Malachite green Monoclonal Antibody (2E10)
GREENER_CHOICE

Catalog No. PIMA549678
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549678 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549678 Supplier Invitrogen™ Supplier No. MA549678
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Malachite green is an organic compound that is used as a dye stuff and controversially as an antimicrobial in aquaculture. Malachite green is traditionally used as a dye for materials such as silk, leather, and paper. Despite its name the dye is not prepared from the mineral malachite, and the name just comes from the similarity of color. Numerous niche applications exploit the intense color of Malachite green. It is used as a biological stain for microscopic analysis of cell biology and tissue samples. In 1992, Canadian authorities determined that eating fish contaminated with malachite green posed a significant health risk. Malachite green was classified a Class II Health Hazard.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Malachite green
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2E10
Concentration 7.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Aniline green; Basic green 4; Diamond green B; Victoria green B
Host Species Mouse
Immunogen Conjugate of MG hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.