Learn More
Invitrogen™ Malachite green Monoclonal Antibody (E0-A4), Biotin

Description
Application notes: Indirect ELISA.
Specifications
Specifications
| Antigen | Malachite green |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | E0-A4 |
| Concentration | 2 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% BSA and 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 |
| Gene Alias | Aniline green; Basic green 4; Diamond green B; Victoria green B |
| Host Species | Mouse |
| Immunogen | Malachite green linked to BSA. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.