Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MALD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144255
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144255 Supplier Invitrogen™ Supplier No. PA5144255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MALD1 Polyclonal Antibody for Western Blot, ELISA

MALD1 (MicroRNA-activated long non-coding RNA 1) is a long non-coding RNA (lncRNA) molecule that has been identified in scientific research. Long non-coding RNAs are RNA molecules that are longer than 200 nucleotides and do not code for proteins. They have been found to play important roles in various cellular processes, including gene regulation, chromatin remodeling, and cellular development. The specific function of MALD1 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. It is worth noting that lncRNAs have been implicated in various biological processes and diseases, including cancer, cardiovascular diseases, and neurological disorders. Therefore, understanding the specific functions and mechanisms of lncRNAs like MALD1 may provide valuable insights into the molecular mechanisms underlying these conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MALD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MALD1
Gene Accession No. P43211
Gene Alias allergen Mal d 1; Allergen Mal d I; Major allergen Mal d 1; MALD1
Gene Symbols MALD1
Host Species Rabbit
Immunogen Recombinant Apple Major allergen Mal d 1 protein (2-159AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103425638
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.