Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Colder Products Company Male Luer Genderless MicroCNX Connector
Shop All Colder Products Company Products
Click to view available options
:
Connector Type: Male
Description
- The MicroCNX line of connectors is engineered specifically for the challenging conditions of biological media transfer in bioprocessing, cell therapy, and gene therapy applications.
- The connector comes in a range of tubing sizes; a luer connection option to create a sterile, closed connection on an existing luer connection; and the ULT version is ideal for assemblies for cryogenic freezing cassettes or connection to PVC tubing.
- Finish: Molded White
- Pressure Range: Up to 43.5 psi, 3 bar
- Free Floating
- Temperature Range: 34°F to 104°F (1°C to 40°C)
- Seal Options: Silicone, platinum-cured, USP Class VI, ADCF
- Non-Valved
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polycarbonate |
| Sterility | Non-sterile |
| Color | White, Blue |
| Product Type | Connector |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction