Learn More
Restek™ Male Pipe to Valco™ Internal Adapter Connector
SDP
Catalog No. 06710527 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Port Type:
1/4 in. NPT Male to 1/16 in. ZDV
1/8 in. NPT Male to 1/16 in. ZDV
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Type
06-710-527 1/8 in. NPT Male to 1/16 in. ZDV
06-710-528 1/4 in. NPT Male to 1/16 in. ZDV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-710-527 Supplier Restek™ Supplier No. 20158
Add to Cart
Add to Cart
Makes a minimum volume connection from a female pipe fitting on a pressure gauge or regulator to a Valco zero-dead-volume fitting.
  • 300 series stainless steel
  • Includes stainless steel ferrules
  • Union Bore: 1.0mm
Specifications
Type Connector
Diameter (Metric) Inner 1 mm
Thread Size 1/8 in. NPT male to 1/16 in. ZDV
Port Type 1/8 in. NPT Male to 1/16 in. ZDV