Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ malE Recombinant Rabbit Monoclonal Antibody (10F2)
GREENER_CHOICE

Catalog No. FEMA550197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550197 Supplier Invitrogen™ Supplier No. MA550197
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

malE Recombinant Monoclonal Antibody for Western Blot, ELISA

Part of the ABC transporter complex MalEFGK involved in maltose/maltodextrin import. Binds maltose and higher maltodextrins such as maltotriose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen malE
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 10F2
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene malE
Gene Accession No. P0AEX9
Gene Alias malE; Maltose-binding periplasmic protein
Gene Symbols malE
Host Species Rabbit
Immunogen A synthesized peptide derived from Escherichia coli malE.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75204178
Target Species Human, Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.